Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011711.1 | NP_035841.1 | 22 | 2784 | ||
GenBank mRNA | AK129473.1 | - | 22 | 2703 | ||
AK139711.1 | - | 22 | 2769 | |||
AK143308.1 | - | 22 | 2684 | |||
AK155331.1 | - | 22 | 2801 | |||
AK163361.1 | - | 22 | 2771 | |||
AK170744.1 | - | 21 | 2614 | |||
BC023797.1 | - | 21 | 2504 | |||
BC027845.1 | - | 6 | 761 | |||
BC085094.1 | - | 22 | 2679 | |||
BC131961.1 | - | 22 | 2660 | 40130238 |