Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166434.2 | NP_001159906.1 | 10 | 1374 | ||
NM_001166435.2 | NP_001159907.1 | 8 | 1105 | |||
NM_001166436.2 | NP_001159908.1 | 8 | 991 | |||
NM_002215.3 | NP_002206.2 | 12 | 1535 | |||
GenBank mRNA | AF116713.2 | - | 3 | 397 | ||
AK292750.1 | - | 12 | 1505 | |||
AK298398.1 | - | 12 | 1534 | |||
AK298423.1 | - | 12 | 1527 | |||
AK298455.1 | - | 10 | 1360 | |||
AK303110.1 | - | 8 | 991 | |||
AK303156.1 | - | 8 | 1105 | |||
AK314198.1 | - | 12 | 1505 | |||
BC069464.1 | - | 12 | 1529 | 7262464 | ||
M18192.1 | - | 3 | 312 | |||
X16260.1 | - | 10 | 1284 | |||
X63652.1 | - | 12 | 1529 |