Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011716.2 | NP_035846.1 | 8 | 2450 | ||
GenBank mRNA | AF084482.1 | - | 8 | 2383 | ||
AJ011971.1 | - | 8 | 2377 | |||
AK144561.1 | - | 8 | 2174 | |||
AK145023.1 | - | 8 | 2422 | |||
AK170180.1 | - | 8 | 2405 | |||
BC046988.1 | - | 8 | 2386 | 6492038 |