Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002216.2 | NP_002207.2 | 13 | 1773 | ||
GenBank mRNA | AK225277.1 | - | 15 | 1636 | ||
AK225329.1 | - | 13 | 1678 | |||
AK291379.1 | - | 13 | 1721 | |||
AK297334.1 | - | 8 | 964 | |||
BC132685.1 | - | 13 | 1660 | 40146707 | ||
CB154170.1 | - | 4 | 546 | |||
M18193.1 | - | 3 | 464 | |||
M33033.1 | - | 3 | 462 | |||
X07173.1 | - | 13 | 1674 |