Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011717.4 | NP_035847.2 | 7, 8 | 2921 | ||
NM_212438.3 | NP_997603.3 | 6, 7 | 2577 | |||
GenBank mRNA | AB012265.1 | - | 11, 12 | 5106 | ||
AB012266.1 | - | 7, 8 | 2615 | |||
AB255388.1 | - | 6, 7 | 2519 | |||
AK004274.1 | - | 1, 2 | 130 | |||
BC053035.1 | - | 9, 10 | 2953 | 6400515 | ||
BG805588.1 | - | 1, 2 | 39 |