Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285792.1 | NP_001272721.1 | 6 | 1116 | ||
NM_001285794.1 | NP_001272723.1 | 5 | 874 | |||
NM_001285795.1 | NP_001272724.1 | 4 | 718 | |||
NM_009519.2 | NP_033545.1 | 4 | 882 | |||
GenBank mRNA | AK031573.1 | - | 4 | 726 | ||
AK077828.1 | - | 5 | 813 | |||
AK138967.1 | - | 5 | 875 | |||
BC125482.1 | - | 4 | 829 | 40129997 | ||
BC125484.1 | - | 4 | 829 | 40129999 | ||
BC144947.1 | - | 4 | 665 | |||
X70800.1 | - | 4 | 889 |