Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144783.2 | NP_659032.3 | 9 | 1624 | ||
GenBank mRNA | BG087452.2 | - | 5 | 423 | ||
DQ537939.1 | - | 9 | 961 | |||
M55512.1 | - | 9 | 1629 |