Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271730.1 | NP_001258659.1 | 6 | 949 | ||
NM_013842.3 | NP_038870.2 | 5 | 975 | |||
GenBank mRNA | AF027963.3 | - | 5 | 649 | ||
AF443192.1 | - | 6 | 949 | |||
AK007724.1 | - | 5 | 636 | |||
AK145241.1 | - | 5 | 635 | |||
AK178083.1 | - | 2 | 145 | |||
AK187998.1 | - | 2 | 145 | |||
AK191275.1 | - | 2 | 145 | |||
AK195676.1 | - | 2 | 145 | |||
AK201525.1 | - | 2 | 134 | |||
AK202632.1 | - | 2 | 163 | |||
AK204892.1 | - | 2 | 171 | |||
AK206863.1 | - | 2 | 146 | |||
AK207925.1 | - | 2 | 160 | |||
AK209182.1 | - | 2 | 145 | |||
AK211765.1 | - | 2 | 160 | |||
AK213043.1 | - | 2 | 160 | |||
AK216856.1 | - | 2 | 145 | |||
AK217319.1 | - | 2 | 145 | |||
AK217907.1 | - | 2 | 145 | |||
AK218355.1 | - | 2 | 145 | |||
AK218387.1 | - | 2 | 144 | |||
AK218413.1 | - | 2 | 145 | |||
AK219822.1 | - | 1 | 174 | |||
BC003496.1 | - | 1 | 105 | |||
BC008153.1 | - | 5 | 590 | |||
BC016079.1 | - | 5 | 869 | 4489182 | ||
BC029197.1 | - | 5 | 630 | 2648085 |