Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271730.1 | NP_001258659.1 | 3 | 690 | ||
NM_013842.3 | NP_038870.2 | 3 | 690 | |||
GenBank mRNA | AF027963.3 | - | 3 | 364 | ||
AF443192.1 | - | 3 | 690 | |||
AK007724.1 | - | 3 | 351 | |||
AK145241.1 | - | 3 | 350 | |||
BC008153.1 | - | 3 | 305 | |||
BC016079.1 | - | 3 | 655 | 4489182 | ||
BC029197.1 | - | 3 | 345 | 2648085 | ||
BG800705.1 | - | 3 | 321 |