Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011723.3 | NP_035853.2 | 26 | 2939 | ||
GenBank mRNA | AK034256.1 | - | 26 | 2887 | ||
AK090058.1 | - | 17 | 2081 | |||
AK134156.1 | - | 12 | 1424 | |||
AK143412.1 | - | 26 | 2888 | |||
AK144700.1 | - | 26 | 2891 | |||
AK160010.1 | - | 23 | 2638 | |||
AK161529.1 | - | 26 | 2890 | |||
AK171642.1 | - | 7 | 781 | |||
BC032077.1 | - | 10 | 1144 | |||
BC065699.1 | - | 17 | 2076 | |||
BC141183.1 | - | 26 | 2914 | 9055795 | ||
X62932.1 | - | 26 | 2873 |