Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018871.3 | NP_061359.2 | 2 | 869 | ||
GenBank mRNA | AB073970.1 | - | 2 | 816 | ||
AF058799.1 | - | 2 | 819 | |||
AK088847.1 | - | 2 | 782 | |||
AK148618.1 | - | 2 | 828 | |||
AK153307.1 | - | 2 | 828 | |||
AK164356.1 | - | 2 | 820 | |||
BC008129.1 | - | 2 | 752 | 3498633 | ||
CT010208.1 | - | 2 | 637 |