Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011739.3 | NP_035869.1 | 2 | 679 | ||
GenBank mRNA | AK008948.1 | - | 3 | 449 | ||
AK012300.1 | - | 2 | 412 | |||
AK012540.1 | - | 3 | 414 | |||
AK145568.1 | - | 3 | 404 | |||
AK146371.1 | - | 3 | 404 | |||
AK150850.1 | - | 3 | 404 | |||
AK151437.1 | - | 3 | 453 | |||
AK151715.1 | - | 3 | 432 | |||
AK151864.1 | - | 3 | 453 | |||
AK159403.1 | - | 3 | 437 | |||
AK159817.1 | - | 3 | 453 | |||
AK163237.1 | - | 3 | 406 | |||
AK167397.1 | - | 3 | 405 | |||
AK182050.1 | - | 2 | 166 | |||
AK190134.1 | - | 2 | 164 | |||
AK192231.1 | - | 2 | 164 | |||
AK193357.1 | - | 2 | 164 | |||
AK196448.1 | - | 2 | 164 | |||
AK201174.1 | - | 2 | 165 | |||
BC003837.1 | - | 2 | 192 | |||
BC080802.1 | - | 3 | 394 | |||
BC085299.1 | - | 3 | 459 | |||
BC090838.1 | - | 2 | 679 | 6403848 | ||
BC106164.1 | - | 2 | 381 | 3582392 | ||
BC110680.1 | - | 2 | 388 | |||
BC132252.1 | - | 2 | 448 | 40130529 | ||
BC132530.1 | - | 2 | 448 | 40130807 | ||
D87662.1 | - | 2 | 371 | |||
U56243.1 | - | 2 | 354 | |||
U57312.1 | - | 2 | 334 |