Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011739.3 | NP_035869.1 | 1 | 590 | ||
GenBank mRNA | AK008948.1 | - | 2 | 360 | ||
AK012540.1 | - | 2 | 325 | |||
AK145568.1 | - | 2 | 315 | |||
AK146371.1 | - | 2 | 315 | |||
AK150850.1 | - | 2 | 315 | |||
AK151437.1 | - | 2 | 364 | |||
AK151715.1 | - | 2 | 343 | |||
AK151864.1 | - | 2 | 364 | |||
AK159403.1 | - | 2 | 348 | |||
AK159817.1 | - | 2 | 364 | |||
AK163237.1 | - | 2 | 317 | |||
AK167397.1 | - | 2 | 316 | |||
AK177592.1 | - | 1 | 75 | |||
AK182050.1 | - | 1 | 77 | |||
AK185231.1 | - | 1 | 77 | |||
AK190134.1 | - | 1 | 75 | |||
AK192231.1 | - | 1 | 75 | |||
AK193357.1 | - | 1 | 75 | |||
AK196448.1 | - | 1 | 75 | |||
AK198944.1 | - | 1 | 75 | |||
AK201174.1 | - | 1 | 75 | |||
BC003837.1 | - | 1 | 103 | |||
BC080802.1 | - | 2 | 305 | |||
BC085299.1 | - | 2 | 370 | |||
BC090838.1 | - | 1 | 590 | 6403848 | ||
BC106164.1 | - | 1 | 292 | 3582392 | ||
BC110680.1 | - | 1 | 298 | |||
BC132252.1 | - | 1 | 359 | 40130529 | ||
BC132530.1 | - | 1 | 359 | 40130807 | ||
D87662.1 | - | 1 | 282 | |||
U56243.1 | - | 1 | 265 | |||
U57312.1 | - | 1 | 245 |