Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011739.3 | NP_035869.1 | 1 | 427 | ||
GenBank mRNA | AK008948.1 | - | 2 | 198 | ||
AK012540.1 | - | 2 | 163 | |||
AK145568.1 | - | 2 | 152 | |||
AK146371.1 | - | 2 | 152 | |||
AK150850.1 | - | 2 | 152 | |||
AK151437.1 | - | 2 | 201 | |||
AK151715.1 | - | 2 | 180 | |||
AK151864.1 | - | 2 | 201 | |||
AK159403.1 | - | 2 | 185 | |||
AK163237.1 | - | 2 | 154 | |||
AK167397.1 | - | 2 | 153 | |||
AK184566.1 | - | 1 | 99 | |||
AK203628.1 | - | 1 | 108 | |||
BC080802.1 | - | 2 | 142 | |||
BC085299.1 | - | 2 | 207 | |||
BC090838.1 | - | 1 | 427 | 6403848 | ||
BC106164.1 | - | 1 | 129 | 3582392 | ||
BC110680.1 | - | 1 | 135 | |||
BC132252.1 | - | 1 | 196 | 40130529 | ||
BC132530.1 | - | 1 | 196 | 40130807 | ||
D87662.1 | - | 1 | 119 | |||
U56243.1 | - | 1 | 102 | |||
U57312.1 | - | 1 | 82 |