Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253805.1 | NP_001240734.1 | 3 | 759 | ||
NM_001253806.1 | NP_001240735.1 | 3 | 484 | |||
NM_001253807.1 | NP_001240736.1 | 2 | 319 | |||
NM_011740.3 | NP_035870.1 | 4 | 956 | |||
GenBank mRNA | AK083368.1 | - | 3 | 487 | ||
AK145657.1 | - | 3 | 418 | |||
AK146800.1 | - | 3 | 475 | |||
AK150381.1 | - | 3 | 487 | |||
AK151900.1 | - | 3 | 448 | |||
AK162099.1 | - | 3 | 439 | |||
AK167128.1 | - | 3 | 456 | |||
BC050891.1 | - | 4 | 662 | 6419584 | ||
BC089334.1 | - | 3 | 387 | 30361690 | ||
CF172373.1 | - | 3 | 484 | |||
D78647.1 | - | 3 | 407 | |||
D83037.1 | - | 3 | 441 | |||
D87660.1 | - | 3 | 489 | |||
U79231.1 | - | 2 | 354 |