Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267623.1 | NP_001254552.1 | 5, 6 | 565 | ||
NM_001324236.1 | NP_001311165.1 | 6, 7 | 691 | |||
NM_001324237.1 | NP_001311166.1 | 7, 8 | 596 | |||
NM_001324238.1 | NP_001311167.1 | 6, 7 | 599 | |||
NM_033453.3 | NP_258412.1 | 7, 8 | 688 | |||
NM_181493.3 | NP_852470.1 | 7, 8 | 637 | |||
NR_052000.1 | 6, 7 | 531 | ||||
NR_052001.2 | 6, 7 | 609 | ||||
NR_052002.1 | 6, 7 | 623 | ||||
XM_006723564.3 | XP_006723627.1 | 8, 9 | 1018 | |||
XM_006723565.3 | XP_006723628.1 | 6, 7 | 895 | |||
GenBank mRNA | AB062127.1 | - | 7, 8 | 557 | ||
AF026816.2 | - | 7, 8 | 547 | |||
AF063607.1 | - | 7, 8 | 572 | |||
AF219116.1 | - | 7, 8 | 600 | |||
AF334680.1 | - | 6, 7 | 480 | |||
BC010138.2 | - | 7, 8 | 532 | 4124624 | ||
BE275997.1 | - | 6, 7 | 566 | |||
BF969192.1 | - | 6, 7 | 531 | |||
BI115811.1 | - | 5, 6 | 401 | |||
BM560812.1 | - | 688 | ||||
BQ049090.1 | - | 6, 7 | 470 | |||
EF199841.1 | - | 7, 8 | 535 | |||
EF213026.1 | - | 7, 8 | 484 | |||
KU178148.1 | - | 7, 8 | 496 | |||
KU178149.1 | - | 5, 6 | 373 |