Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253805.1 | NP_001240734.1 | 5 | 1051 | ||
NM_001253806.1 | NP_001240735.1 | 5 | 776 | |||
NM_001253807.1 | NP_001240736.1 | 4 | 611 | |||
NM_011740.3 | NP_035870.1 | 6 | 1248 | |||
GenBank mRNA | AK083368.1 | - | 5 | 779 | ||
AK145657.1 | - | 5 | 710 | |||
AK146800.1 | - | 5 | 767 | |||
AK150381.1 | - | 5 | 779 | |||
AK151506.1 | - | 3 | 247 | |||
AK151900.1 | - | 5 | 740 | |||
AK162099.1 | - | 5 | 731 | |||
AK167128.1 | - | 5 | 748 | |||
AK187117.1 | - | 3 | 333 | |||
AK201135.1 | - | 3 | 277 | |||
BC050891.1 | - | 6 | 954 | 6419584 | ||
BC089334.1 | - | 5 | 679 | 30361690 | ||
D78647.1 | - | 5 | 699 | |||
D83037.1 | - | 5 | 733 | |||
U79231.1 | - | 4 | 646 |