Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267623.1 | NP_001254552.1 | 5 | 538 | ||
NM_001324236.1 | NP_001311165.1 | 6 | 664 | |||
NM_001324237.1 | NP_001311166.1 | 7 | 569 | |||
NM_001324238.1 | NP_001311167.1 | 6 | 572 | |||
NM_033453.3 | NP_258412.1 | 7 | 661 | |||
NM_181493.3 | NP_852470.1 | 7 | 610 | |||
NR_052000.1 | 6 | 504 | ||||
NR_052001.2 | 6 | 582 | ||||
NR_052002.1 | 6 | 596 | ||||
XM_006723564.3 | XP_006723627.1 | 8 | 991 | |||
XM_006723565.3 | XP_006723628.1 | 6 | 868 | |||
GenBank mRNA | AB062127.1 | - | 7 | 530 | ||
AF026816.2 | - | 7 | 520 | |||
AF219116.1 | - | 7 | 573 | |||
AF334680.1 | - | 6 | 453 | |||
BC010138.2 | - | 7 | 505 | 4124624 | ||
BE275997.1 | - | 6 | 539 | |||
BF969192.1 | - | 6 | 504 | |||
BI115811.1 | - | 5 | 374 | |||
BM560812.1 | - | 661 | ||||
BQ049090.1 | - | 6 | 443 | |||
EF199841.1 | - | 7 | 508 | |||
EF213026.1 | - | 7 | 457 | |||
KU178148.1 | - | 7 | 469 | |||
KU178149.1 | - | 5 | 346 |