Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfa-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029509.1 | - | 1 | 1134 |
Gene Symbol: Zfx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC144979.1 | - | 2, 3 | 669 | 9053527 |