Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021263.1 | XP_016876752.1 | 2, 3 | 157 | ||
GenBank mRNA | AF279372.1 | - | 2, 3 | 177 | ||
AK293515.1 | - | 2, 3 | 232 | |||
BC003622.2 | - | 3, 4 | 337 | 3010239 | ||
BC007428.2 | - | 3, 4 | 337 | 3010239 | ||
BC018192.1 | - | 3, 4 | 286 | 3867269 | ||
BQ434653.1 | - | 3, 4 | 365 | |||
BX420122.2 | - | 3, 4 | 401 | |||
CB997740.1 | - | 255 | ||||
U51336.1 | - | 3, 4 | 230 |