Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009551.5 | NP_033577.1 | 2 | 569 | ||
GenBank mRNA | AF062071.1 | - | 1 | 161 | ||
AK076397.1 | - | 2 | 570 | |||
AK131911.1 | - | 2 | 565 | |||
AK211340.1 | - | 1 | 110 | |||
AK218278.1 | - | 1 | 110 | |||
BC048232.1 | - | 1 | 550 | |||
BC061693.1 | - | 1 | 490 | |||
BC080270.1 | - | 2 | 551 | |||
BC107566.1 | - | 2 | 133 | 5389770 | ||
BC119124.1 | - | 2 | 570 | 8733873 | ||
BC119126.1 | - | 2 | 570 | 8733875 |