Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011753.3 | NP_035883.2 | 10 | 1869 | ||
GenBank mRNA | AB010345.1 | - | 1 | 225 | ||
AK029298.1 | - | 8 | 2134 | |||
AK082002.1 | - | 10 | 1703 | |||
AK141740.1 | - | 9 | 2232 | |||
BC051056.1 | - | 3 | 947 |