Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037707.1 | NP_001032796.1 | 5 | 742 | ||
NM_001285797.1 | NP_001272726.1 | 5 | 1014 | |||
NM_001285798.1 | NP_001272727.1 | 5 | 968 | |||
NM_011754.2 | NP_035884.2 | 5 | 737 | |||
GenBank mRNA | AK028500.1 | - | 5 | 969 | ||
AK135293.1 | - | 5 | 1018 | |||
AK147732.1 | - | 5 | 732 | |||
BC059037.1 | - | 3 | 681 | 6812360 | ||
BY740816.1 | - | 5 | 732 |