Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044703.2 | NP_001038168.1 | 4 | 1390 | ||
NM_001044704.2 | NP_001038169.1 | 5 | 1456 | |||
NM_001044705.2 | NP_001038170.1 | 4 | 1371 | |||
NM_011757.3 | NP_035887.2 | 5 | 1475 | |||
GenBank mRNA | AK146776.1 | - | 6 | 1411 | ||
AK146875.1 | - | 5 | 1436 | |||
BC010591.1 | - | 4 | 1318 | 3981599 | ||
D10630.1 | - | 4 | 1309 | |||
X63747.1 | - | 4 | 2166 |