Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044703.2 | NP_001038168.1 | 4 | 1711 | ||
NM_001044704.2 | NP_001038169.1 | 5 | 1777 | |||
NM_001044705.2 | NP_001038170.1 | 4 | 1692 | |||
NM_011757.3 | NP_035887.2 | 5 | 1796 | |||
GenBank mRNA | AK146776.1 | - | 6 | 1732 | ||
AK146875.1 | - | 5 | 1757 | |||
BC010591.1 | - | 4 | 1639 | 3981599 | ||
D10630.1 | - | 4 | 1630 | |||
U41671.1 | - | 5 | 1724 |