Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002221.3 | NP_002212.3 | 2 | 1100 | ||
XM_005273120.2 | XP_005273177.1 | 2 | 1252 | |||
XM_017001211.1 | XP_016856700.1 | 2 | 1252 | |||
GenBank mRNA | AJ242780.1 | - | 1 | 817 | ||
BC015009.1 | - | 2 | 1100 | 4862726 | ||
Y18024.2 | - | 1 | 760 |