Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099952.2 | NP_001093422.2 | 40 | 5458 | ||
NM_001168272.1 | NP_001161744.1 | 42 | 5557 | |||
NM_002222.5 | NP_002213.5 | 39 | 5413 | |||
XM_005265109.2 | XP_005265166.1 | 41 | 5533 | |||
XM_005265110.2 | XP_005265167.1 | 40 | 5485 | |||
XM_006713131.2 | XP_006713194.1 | 41 | 5536 | |||
XM_011533681.1 | XP_011531983.1 | 43 | 5605 | |||
XM_011533682.2 | XP_011531984.1 | 43 | 5531 | |||
XM_011533683.2 | XP_011531985.1 | 43 | 5602 | |||
XM_011533684.1 | XP_011531986.1 | 43 | 5578 | |||
XM_011533685.1 | XP_011531987.1 | 42 | 5572 | |||
XM_011533686.1 | XP_011531988.1 | 42 | 5569 | |||
XM_011533687.1 | XP_011531989.1 | 42 | 5560 | |||
XM_011533688.1 | XP_011531990.1 | 42 | 5533 | |||
XM_011533690.1 | XP_011531992.1 | 43 | 5605 | |||
XM_011533691.1 | XP_011531993.1 | 43 | 5605 | |||
XM_011533692.2 | XP_011531994.1 | 43 | 5605 | |||
XM_017006357.1 | XP_016861846.1 | 43 | 5602 | |||
XM_017006358.1 | XP_016861847.1 | 43 | 5602 | |||
GenBank mRNA | AB208868.1 | - | 20 | 2329 | ||
D26070.1 | - | 39 | 5319 | |||
L38019.2 | - | 40 | 5355 | |||
S82269.1 | - | 4 | 466 | |||
U23850.1 | - | 40 | 5207 |