Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011762.3 | NP_035892.2 | 5 | 1286 | ||
GenBank mRNA | AK142951.1 | - | 5 | 1263 |
Gene Symbol: Gm7452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC127053.1 | - | 1 | 1062 |