Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009567.4 | NP_033593.1 | 4 | 849 | ||
GenBank mRNA | AK028905.1 | - | 4 | 687 | ||
AK040325.1 | - | 4 | 780 | |||
AK132229.1 | - | 4 | 850 | |||
AK146095.1 | - | 4 | 840 | |||
AK165630.1 | - | 4 | 810 | |||
BC003776.1 | - | 572 | 3496816 | |||
BC061487.1 | - | 3 | 630 | 5349153 | ||
U46186.1 | - | 4 | 841 |