Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009567.4 | NP_033593.1 | 4 | 1706 | ||
GenBank mRNA | AK028905.1 | - | 4 | 1544 | ||
AK040325.1 | - | 4 | 1637 | |||
AK132229.1 | - | 4 | 1707 | |||
AK146095.1 | - | 4 | 1697 | |||
AK165630.1 | - | 4 | 1661 | |||
BC003776.1 | - | 1429 | 3496816 | |||
BC061487.1 | - | 3 | 1487 | 5349153 | ||
U46186.1 | - | 4 | 1698 |