Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167944.1 | NP_001161416.1 | 4 | 2600 | ||
NM_016683.2 | NP_057892.2 | 6 | 2819 | |||
GenBank mRNA | AB221634.1 | - | 6 | 2083 | ||
BC060248.1 | - | 4 | 2495 | 5715760 | ||
U62907.1 | - | 6 | 2143 |