Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009574.3 | NP_033600.3 | 3 | 1965 | ||
GenBank mRNA | AK135125.1 | - | 1 | 77 | ||
D70848.2 | - | 3 | 1918 |