Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009576.2 | NP_033602.2 | 3 | 931 | ||
GenBank mRNA | AK028305.1 | - | 1 | 734 | ||
AK035908.1 | - | 1 | 735 | |||
AK046913.1 | - | 3 | 932 | |||
AK163477.1 | - | 3 | 2005 | |||
D78174.1 | - | 1 | 1184 |