Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036833.1 | - | 6 | 940 | ||
AK040268.1 | - | 5 | 673 | |||
AK077815.1 | - | 6 | 1237 | |||
AK154175.1 | - | 6 | 962 | |||
AK220235.1 | - | 6 | 906 | |||
AY536269.1 | - | 3 | 250 | |||
BC138789.1 | - | 5 | 471 | 8861811 | ||
L03547.1 | - | 5 | 456 | |||
S74708.1 | - | 3 | 517 |