Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290993.1 | NP_001277922.1 | 5, 6 | 1238 | ||
NM_011774.3 | NP_035904.2 | 6, 7 | 1385 | |||
GenBank mRNA | AF003747.1 | - | 6, 7 | 1186 | ||
AK134245.1 | - | 6, 7 | 1363 | |||
AK166106.1 | - | 5, 6 | 1240 | |||
BC117997.1 | - | 6, 7 | 1243 | 40095367 |