Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289617.1 | NP_001276546.1 | 5 | 1288 | ||
NM_001289618.1 | NP_001276547.1 | 4 | 1195 | |||
NM_001289619.1 | NP_001276548.1 | 4 | 1195 | |||
NM_011777.3 | NP_035907.1 | 5 | 1288 | |||
GenBank mRNA | AK031415.1 | - | 5 | 1086 | ||
AK035743.1 | - | 5 | 1059 | |||
AK076491.1 | - | 5 | 1059 | |||
AK090110.1 | - | 5 | 1055 | |||
AK147812.1 | - | 5 | 1289 | |||
AK161709.1 | - | 4 | 990 | |||
AK165290.1 | - | 3 | 1129 | |||
AK170571.1 | - | 5 | 1062 | |||
AK190613.1 | - | 1 | 117 | |||
BC054775.1 | - | 4 | 1154 | 6808149 | ||
X99063.1 | - | 5 | 1246 | |||
Y07711.1 | - | 4 | 996 |