Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011778.2 | NP_035908.1 | 9 | 1110 | ||
GenBank mRNA | AF143956.1 | - | 9 | 1117 | ||
AK008947.1 | - | 5 | 550 | |||
AK149639.1 | - | 9 | 1120 | |||
AK177038.1 | - | 2 | 158 | |||
AK184918.1 | - | 2 | 158 | |||
AK195135.1 | - | 2 | 158 | |||
AK197317.1 | - | 4 | 371 | |||
AK204983.1 | - | 2 | 158 | |||
AK205268.1 | - | 4 | 314 | |||
AK207237.1 | - | 2 | 158 | |||
AK212635.1 | - | 2 | 158 | |||
AK215642.1 | - | 2 | 158 | |||
BC029326.1 | - | 9 | 1069 | |||
BC131944.1 | - | 9 | 1057 | 40130221 | ||
BC132617.1 | - | 9 | 1057 | 40130894 |