Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011783.2 | NP_035913.1 | 3 | 234 | ||
GenBank mRNA | AB016592.1 | - | 3 | 233 | ||
AF044262.1 | - | 3 | 234 | |||
AK007677.1 | - | 3 | 253 | |||
BC013334.1 | - | 3 | 252 | 4218549 |