Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011784.3 | NP_035914.1 | 1 | 1018 | ||
GenBank mRNA | AJ007612.1 | - | 1 | 745 | ||
AK050848.1 | - | 1 | 344 | |||
AK052383.1 | - | 1 | 923 | |||
AK075706.1 | - | 1 | 956 | |||
AK078374.1 | - | 1 | 903 | |||
AK087138.1 | - | 1 | 963 | |||
AK133580.1 | - | 1 | 1019 | |||
AK157582.1 | - | 1 | 983 | |||
BC039224.1 | - | 1 | 987 | 4457726 |
Set Membership: |
G-protein coupled receptor |