Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ash2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080793.2 | NP_001074262.1 | 10 | 1031 | ||
NM_001286207.1 | NP_001273136.1 | 10 | 1004 | |||
NM_011791.3 | NP_035921.2 | 10 | 1121 | |||
GenBank mRNA | AB020983.2 | - | 10 | 1103 | ||
AK087934.1 | - | 10 | 1096 | |||
AK132471.1 | - | 9 | 1027 | |||
AK145576.1 | - | 10 | 996 | |||
AK145790.1 | - | 10 | 1008 | |||
AK145967.1 | - | 10 | 975 | |||
AK146254.1 | - | 10 | 1033 | |||
AK146599.1 | - | 10 | 1005 | |||
AK146756.1 | - | 10 | 996 | |||
AK146938.1 | - | 10 | 1098 | |||
BC012957.1 | - | 10 | 1108 | 4211433 |