Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038231.2 | NP_001033320.1 | 3 | 631 | ||
NM_001286608.1 | NP_001273537.1 | 3 | 973 | |||
NM_011793.3 | NP_035923.1 | 3 | 839 | |||
GenBank mRNA | AB025349.1 | - | 3 | 571 | ||
AK003025.1 | - | 3 | 615 | |||
AK004212.1 | - | 3 | 598 | |||
AK010257.1 | - | 3 | 617 | |||
AK011079.1 | - | 3 | 620 | |||
AK011100.1 | - | 3 | 618 | |||
AK012588.1 | - | 3 | 601 | |||
AK088896.1 | - | 3 | 595 | |||
AK168039.1 | - | 3 | 620 | |||
AK185753.1 | - | 1 | 273 | |||
BC003709.1 | - | 3 | 608 | 3599828 | ||
CK780850.1 | - | 3 | 603 |