Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013706.2 | NP_038734.1 | 1, 2 | 136 | ||
GenBank mRNA | AK088161.1 | - | 1, 2 | 103 | ||
AK155728.1 | - | 1, 2 | 137 | |||
BC027495.1 | - | 1, 2 | 86 | 1396480 | ||
M55561.1 | - | 1, 2 | 125 | |||
X76696.1 | - | 1, 2 | 112 |