Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000215.3 | NP_000206.2 | 19 | 2763 | ||
XM_005259896.3 | XP_005259953.1 | 19 | 2802 | |||
GenBank mRNA | U09607.1 | - | 18 | 2758 | ||
U31601.1 | - | 19 | 2758 |