Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011809.3 | NP_035939.3 | 10 | 1507 | ||
GenBank mRNA | AK014602.2 | - | 10 | 1429 | ||
AK030537.1 | - | 6 | 809 | |||
AK038001.1 | - | 1 | 1268 | |||
AK038348.1 | - | 10 | 1494 | |||
AK128933.1 | - | 11 | 1381 | |||
AK143687.1 | - | 10 | 1459 | |||
AK145852.1 | - | 10 | 1343 | |||
AK150175.1 | - | 10 | 1436 | |||
AK151315.1 | - | 10 | 1458 | |||
AK152750.1 | - | 10 | 1461 | |||
AK153339.1 | - | 10 | 1459 | |||
AK153478.1 | - | 10 | 1459 | |||
BC005486.1 | - | 10 | 1428 | 3486021 | ||
BC005504.1 | - | 10 | 1368 | 3490813 | ||
CT010235.1 | - | 9 | 1269 | |||
J04103.1 | - | 10 | 1406 |