Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011827.3 | NP_035957.2 | 4 | 431 | ||
GenBank mRNA | AF172930.1 | - | 4 | 431 | ||
AK008005.1 | - | 4 | 430 | |||
BC069220.1 | - | 355 | 640698 |