Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206924.2 | NP_996807.1 | 5 | 734 | ||
GenBank mRNA | AB016490.1 | - | 5 | 679 | ||
AK004469.1 | - | 5 | 646 | |||
AK089048.1 | - | 5 | 646 | |||
AK146337.1 | - | 5 | 735 | |||
AK215601.1 | - | 3 | 248 | |||
BC008139.1 | - | 5 | 698 | 3582930 | ||
BG077727.2 | - | 4 | 300 | |||
BY617278.1 | - | 1 | 46 |