Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011836.3 | NP_035966.2 | 22 | 3846 | ||
GenBank mRNA | AF079520.1 | - | 22 | 3869 | ||
AF083372.1 | - | 22 | 3783 | |||
AK132948.1 | - | 19 | 2833 | |||
BC005535.1 | - | 5 | 591 | |||
BC096366.1 | - | 22 | 3846 | 2865309 |