Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011839.3 | NP_035969.2 | 1 | 1542 | ||
GenBank mRNA | AF126427.1 | - | 1 | 1883 | ||
AF149877.1 | - | 1 | 1284 | |||
AK053645.1 | - | 1 | 1543 | |||
AK144373.1 | - | 1 | 1712 | |||
BC046628.1 | - | 1 | 1508 | 6493460 | ||
BC058730.1 | - | 1 | 1648 |