Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011842.3 | NP_035972.3 | 10 | 1378 | ||
GenBank mRNA | AF159259.1 | - | 10 | 1253 | ||
AK077548.1 | - | 10 | 1360 | |||
AK088158.1 | - | 10 | 1291 | |||
AK147099.1 | - | 10 | 1357 | |||
AK149834.1 | - | 10 | 1424 | |||
AK157717.1 | - | 10 | 1282 | |||
AK159979.1 | - | 10 | 1354 | |||
AK182140.1 | - | 1 | 73 | |||
AK186417.1 | - | 4 | 383 | |||
BC079847.1 | - | 10 | 1378 | 6419411 |