Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011843.2 | NP_035973.1 | 9 | 1123 | ||
GenBank mRNA | AK044656.1 | - | 9 | 1133 | ||
AK080094.1 | - | 9 | 1136 | |||
AK088421.1 | - | 9 | 1136 | |||
AK152482.1 | - | 9 | 1136 | |||
AK152554.1 | - | 9 | 1136 | |||
BC011482.1 | - | 9 | 1123 | 4165946 |